Anti-SLC25A24 Rabbit Recombinant Polyclonal Antibody
Catalog # 77829-657
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Calcium-binding mitochondrial carrier protein SCaMC-1 protein (1-197AA)
- Antigen symbol:SLC25A24
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q6NUK1
- Antigen synonyms:SLC25A24|APC1|MCSC1|SCAMC1|Calcium-binding mitochondrial carrier protein SCaMC-1|Mitochondrial ATP-Mg/Pi carrier protein 1|Mitochondrial Ca(2+)-dependent solute carrier protein 1|Small calcium-binding mitochondrial carrier protein 1|Solute carrier family 25 member 24
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: SLC25A24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Rat,Mouse