Anti-APP Rabbit Recombinant Polyclonal Antibody
Catalog # 77810-688
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Amyloid-beta A4 protein (18-270AA)
- Antigen symbol:APP
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P05067
- Antigen synonyms:A4| A4_HUMAN| AAA| ABETA| ABPP| AICD-50| AICD-57| AICD-59| AID(50)| AID(57)| AID(59)| Alzheimer disease amyloid protein| Amyloid intracellular domain 50| Amyloid intracellular domain 57| Amyloid intracellular domain 59| APP| APPI| Beta amyloid protein 42| Beta APP42| Beta-APP40| Beta-APP42| C31| Cerebral vascular amyloid peptide| CVAP| Gamma-CTF(50)| Gamma-CTF(57)| Gamma-CTF(59)| PN-II| PreA4| Protease nexin-II| S-APP-alpha| S-APP-beta
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: APP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse