Anti-AMFR Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AMFR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77815-961
- 77815-962
- Antigen symbol:
- AMFR
- AMFR
- Antigen name:
- E3 ubiquitin-protein ligase AMFR protein (485-643AA)
- E3 ubiquitin-protein ligase AMFR protein (485-643AA)
- Antigen synonyms:
- AMF receptor|AMF receptor isoform 1|AMF receptor isoform 2|AMFR|AMFR2_HUMAN|Autocrine motility factor receptor|Autocrine motility factor receptor precursor|isoform 1|isoform 2|E3 ubiquitin-protein ligase AMFR|gp78|RING finger protein 45|RNF 45|RNF45
- AMF receptor|AMF receptor isoform 1|AMF receptor isoform 2|AMFR|AMFR2_HUMAN|Autocrine motility factor receptor|Autocrine motility factor receptor precursor|isoform 1|isoform 2|E3 ubiquitin-protein ligase AMFR|gp78|RING finger protein 45|RNF 45|RNF45
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKV5
- Q9UKV5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together