Anti-UBE2O Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: UBE2O
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-002
- 77837-003
- Antigen symbol:
- UBE2O
- UBE2O
- Antigen name:
- (E3-independent) E2 ubiquitin-conjugating enzyme protein (10-277AA)
- (E3-independent) E2 ubiquitin-conjugating enzyme protein (10-277AA)
- Antigen synonyms:
- E2 230K|EC=6.3.2.19|FLJ12878|KIAA1734|Likely Ortholog of Mouse Ubiquitin Conjugating Enzyme|RP23 193A16.5|UBE2O|UBE2O_HUMAN|Ubiquitin carrier protein O|Ubiquitin conjugating enzyme E2 O|Ubiquitin conjugating enzyme E2 of 230 kDa|ubiquitin conjugating enzyme E2O|Ubiquitin protein ligase O|Ubiquitin-conjugating enzyme E2 O|Ubiquitin-conjugating enzyme E2 of 230 kDa|Ubiquitin-conjugating enzyme E2-230K|Ubiquitin-protein ligase O
- E2 230K|EC=6.3.2.19|FLJ12878|KIAA1734|Likely Ortholog of Mouse Ubiquitin Conjugating Enzyme|RP23 193A16.5|UBE2O|UBE2O_HUMAN|Ubiquitin carrier protein O|Ubiquitin conjugating enzyme E2 O|Ubiquitin conjugating enzyme E2 of 230 kDa|ubiquitin conjugating enzyme E2O|Ubiquitin protein ligase O|Ubiquitin-conjugating enzyme E2 O|Ubiquitin-conjugating enzyme E2 of 230 kDa|Ubiquitin-conjugating enzyme E2-230K|Ubiquitin-protein ligase O
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C0C9
- Q9C0C9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together