Anti-HESX1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: HESX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-868
- 77837-869
- Antigen symbol:
- HESX1
- HESX1
- Antigen name:
- Homeobox expressed in ES cells 1 protein (1-185AA)
- Homeobox expressed in ES cells 1 protein (1-185AA)
- Antigen synonyms:
- HESX1| HANF| Homeobox expressed in ES cells 1| Homeobox protein ANF| hAnf
- HESX1| HANF| Homeobox expressed in ES cells 1| Homeobox protein ANF| hAnf
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBX0
- Q9UBX0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together