Anti-ST3GAL2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ST3GAL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-299
- 77823-298
- Antigen symbol:
- ST3GAL2
- ST3GAL2
- Antigen name:
- CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2 protein (28-210AA)
- CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2 protein (28-210AA)
- Antigen synonyms:
- 3-GalNAc-alpha-2|3-sialyltransferase 2|3-sialyltransferase|3-ST 2|Alpha 2|Alpha 2,3-ST|Beta-galactoside alpha-2|Beta-galactoside alpha-2,3-sialyltransferase|CMP N-acetylneuraminate beta galactosamide alpha 2,3 sialyltransferase|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2|Gal NAc6S|Gal-beta-1|Gal-NAc6S|SIA4B_HUMAN|sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase)|Sialyltransferase 4B|SIAT4-B|SIAT4B|ST3 beta-galactoside alpha-2,3-sialyltransferase 2|ST3Gal II|ST3GAL2|ST3GalA.2|ST3GalII
- 3-GalNAc-alpha-2|3-sialyltransferase 2|3-sialyltransferase|3-ST 2|Alpha 2|Alpha 2,3-ST|Beta-galactoside alpha-2|Beta-galactoside alpha-2,3-sialyltransferase|CMP N-acetylneuraminate beta galactosamide alpha 2,3 sialyltransferase|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2|Gal NAc6S|Gal-beta-1|Gal-NAc6S|SIA4B_HUMAN|sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase)|Sialyltransferase 4B|SIAT4-B|SIAT4B|ST3 beta-galactoside alpha-2,3-sialyltransferase 2|ST3Gal II|ST3GAL2|ST3GalA.2|ST3GalII
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16842
- Q16842
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together