Anti-CHMP5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHMP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-055
- 77840-056
- Antigen symbol:
- CHMP5
- CHMP5
- Antigen name:
- Charged multivesicular body protein 5 protein (1-219AA)
- Charged multivesicular body protein 5 protein (1-219AA)
- Antigen synonyms:
- apoptosis-related protein PNAS-2| C9orf83| CGI 34| Charged multivesicular body protein 5| CHMP 5| CHMP family| member 5| chmp5| CHMP5_HUMAN| Chromatin modifying protein 5| Chromatin-modifying protein 5| Chromosome 9 open reading frame 83| HGNC:26942| HSPC177| hVps60| PNAS 2| SNF7 domain containing protein 2| SNF7 domain-containing protein 2| SNF7DC2| Vacuolar protein sorting 60| Vacuolar protein sorting-associated protein 60| Vps60
- apoptosis-related protein PNAS-2| C9orf83| CGI 34| Charged multivesicular body protein 5| CHMP 5| CHMP family| member 5| chmp5| CHMP5_HUMAN| Chromatin modifying protein 5| Chromatin-modifying protein 5| Chromosome 9 open reading frame 83| HGNC:26942| HSPC177| hVps60| PNAS 2| SNF7 domain containing protein 2| SNF7 domain-containing protein 2| SNF7DC2| Vacuolar protein sorting 60| Vacuolar protein sorting-associated protein 60| Vps60
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZZ3
- Q9NZZ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together