Anti-SERTAD1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SERTAD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-431
- 77841-432
- Antigen symbol:
- SERTAD1
- SERTAD1
- Antigen name:
- SERTA domain-containing protein 1 protein (1-236AA)
- SERTA domain-containing protein 1 protein (1-236AA)
- Antigen synonyms:
- CDK4 binding protein p34SEI| CDK4 binding protein p34SEI1| CDK4-binding protein p34SEI1| SEI-1| SEI1| SERTA domain containing 1| SERTA domain-containing protein 1| | SRTD1_HUMAN| Transcriptional regulator interacting with the PHD-bromodomain 1| TRIP Br1| TRIP-Br1
- CDK4 binding protein p34SEI| CDK4 binding protein p34SEI1| CDK4-binding protein p34SEI1| SEI-1| SEI1| SERTA domain containing 1| SERTA domain-containing protein 1| | SRTD1_HUMAN| Transcriptional regulator interacting with the PHD-bromodomain 1| TRIP Br1| TRIP-Br1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHV2
- Q9UHV2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together