Order Entry
Canada
ContactUsLinkComponent
 
Anti-PDE11A Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
 
undefined
Anti-PDE11A Rabbit Recombinant Polyclonal Antibody (FITC)
New Product

 

  • Catalog No:
  • 77839-584
  • 77839-583
  • Antigen symbol:
  • PDE11A
  • PDE11A
  • Antigen name:
  • Dual 3\',5\'-cyclic-AMP and -GMP phosphodiesterase 11A protein (42-190AA)
  • Dual 3\',5\'-cyclic-AMP and -GMP phosphodiesterase 11A protein (42-190AA)
  • Antigen synonyms:
  • 5''-cyclic-AMP and -GMP phosphodiesterase 11A|cAMP and cGMP phosphodiesterase 11A|cGMP phosphodiesterase 11A|Cyclic nucleotide phosphodiesterase 11A|Dual 3''|Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A|pde11|PDE11_HUMAN|pde11a|PDE11A1|pde11a2|Phosphodiesterase 11A|PPNAD2
  • 5''-cyclic-AMP and -GMP phosphodiesterase 11A|cAMP and cGMP phosphodiesterase 11A|cGMP phosphodiesterase 11A|Cyclic nucleotide phosphodiesterase 11A|Dual 3''|Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A|pde11|PDE11_HUMAN|pde11a|PDE11A1|pde11a2|Phosphodiesterase 11A|PPNAD2
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9HCR9
  • Q9HCR9
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together