Anti-SFRP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SFRP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-315
- 77799-316
- Antigen symbol:
- SFRP1
- SFRP1
- Antigen name:
- Secreted frizzled-related protein 1 protein (170-240AA)
- Secreted frizzled-related protein 1 protein (170-240AA)
- Antigen synonyms:
- Frizzled related protein 1|FRP 1|FRP|FRP-1|FRP1|FrzA|SARP 2|SARP-2|SARP2|Secreted apoptosis related protein 2|Secreted apoptosis-related protein 2|Secreted frizzled related protein 1|Secreted frizzled related protein|Secreted frizzled-related protein 1|SFRP 1|sFRP-1|SFRP1|SFRP1_HUMAN
- Frizzled related protein 1|FRP 1|FRP|FRP-1|FRP1|FrzA|SARP 2|SARP-2|SARP2|Secreted apoptosis related protein 2|Secreted apoptosis-related protein 2|Secreted frizzled related protein 1|Secreted frizzled related protein|Secreted frizzled-related protein 1|SFRP 1|sFRP-1|SFRP1|SFRP1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N474
- Q8N474
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together