Anti-CTF1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CTF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-603
- 77824-604
- Antigen symbol:
- CTF1
- CTF1
- Antigen name:
- Cardiotrophin-1 protein (1-201AA)
- Cardiotrophin-1 protein (1-201AA)
- Antigen synonyms:
- Cardiophin 1|Cardiophin1|Cardiotrophin I|Cardiotrophin-1|Cardiotrophin1|CardiotrophinI|CT 1|CT-1|CT1|CTF 1|Ctf1|CTF1_HUMAN
- Cardiophin 1|Cardiophin1|Cardiotrophin I|Cardiotrophin-1|Cardiotrophin1|CardiotrophinI|CT 1|CT-1|CT1|CTF 1|Ctf1|CTF1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16619
- Q16619
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together