Order Entry
Canada
ContactUsLinkComponent
Anti-SEPT3 Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-SEPT3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SEPT3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77799-058
  • 77799-057
  • Antigen symbol:
  • SEPT3
  • SEPT3
  • Antigen name:
  • Neuronal-specific septin-3 protein (238-358AA)
  • Neuronal-specific septin-3 protein (238-358AA)
  • Antigen synonyms:
  • bK250D10.3|MGC133218|Neuronal specific septin 3|Neuronal-specific septin-3|OTTHUMP00000028742|SEP 3|SEP3|SEPT 3|sept3|SEPT3 protein|SEPT3_HUMAN|Septin3
  • bK250D10.3|MGC133218|Neuronal specific septin 3|Neuronal-specific septin-3|OTTHUMP00000028742|SEP 3|SEP3|SEPT 3|sept3|SEPT3 protein|SEPT3_HUMAN|Septin3
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UH03
  • Q9UH03
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together