Anti-SLURP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SLURP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-240
- 77800-241
- Antigen symbol:
- SLURP1
- SLURP1
- Antigen name:
- Secreted Ly-6/uPAR-related protein 1 protein (1-103AA)
- Secreted Ly-6/uPAR-related protein 1 protein (1-103AA)
- Antigen synonyms:
- SLURP1|ARS|Secreted Ly-6/uPAR-related protein 1|SLURP-1|ARS component B|ARS(component B)-81/S|Anti-neoplastic urinary protein|ANUP
- SLURP1|ARS|Secreted Ly-6/uPAR-related protein 1|SLURP-1|ARS component B|ARS(component B)-81/S|Anti-neoplastic urinary protein|ANUP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55000
- P55000
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together