Anti-SPAM1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPAM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-810
- 77800-809
- Antigen symbol:
- SPAM1
- SPAM1
- Antigen name:
- Hyaluronidase PH-20 protein (36-490AA)
- Hyaluronidase PH-20 protein (36-490AA)
- Antigen synonyms:
- epididymis secretory sperm binding protein Li 96n|HEL-S-96n|HYA1|HYAL PH-20|HYAL PH20|Hyal-PH20|HYAL1|HYAL3|HYAL5|HYALP_HUMAN|Hyaluronidase PH-20|Hyaluronoglucosaminidase PH-20|MGC26532|PH-20 Hyaluronidase|PH20|PH20 Hyaluronidase|SPAG15|SPAM-1|Spam1|sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding)|Sperm adhesion molecule 1|Sperm surface protein PH-20|Sperm surface protein PH20|zona pellucida binding
- epididymis secretory sperm binding protein Li 96n|HEL-S-96n|HYA1|HYAL PH-20|HYAL PH20|Hyal-PH20|HYAL1|HYAL3|HYAL5|HYALP_HUMAN|Hyaluronidase PH-20|Hyaluronoglucosaminidase PH-20|MGC26532|PH-20 Hyaluronidase|PH20|PH20 Hyaluronidase|SPAG15|SPAM-1|Spam1|sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding)|Sperm adhesion molecule 1|Sperm surface protein PH-20|Sperm surface protein PH20|zona pellucida binding
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P38567
- P38567
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together