Anti-XPO7 Rabbit Polyclonal Antibody
Catalog # 77804-758
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Ran-binding protein 16
- Antigen symbol:XPO7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UIA9
- Antigen synonyms:XPO7|KIAA0745|RANBP16|Exportin-7|Exp7|Ran-binding protein 16
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: XPO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat