Anti-PLA1A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLA1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-276
- 77794-277
- Antigen symbol:
- PLA1A
- PLA1A
- Antigen name:
- Phospholipase A1 member A protein (293-398AA)
- Phospholipase A1 member A protein (293-398AA)
- Antigen synonyms:
- AA986889|NMD|OTTHUMP00000215248|OTTHUMP00000215249|OTTHUMP00000215250|Phosphatidylserine specific phospholipase A1|Phosphatidylserine specific phospholipase A1alpha|Phosphatidylserine-specific phospholipase A1|Phospholipase A1 member A|pla1a|PLA1A_HUMAN|PS PLA1|PS-PLA1|PSPLA 1|PSPLA1
- AA986889|NMD|OTTHUMP00000215248|OTTHUMP00000215249|OTTHUMP00000215250|Phosphatidylserine specific phospholipase A1|Phosphatidylserine specific phospholipase A1alpha|Phosphatidylserine-specific phospholipase A1|Phospholipase A1 member A|pla1a|PLA1A_HUMAN|PS PLA1|PS-PLA1|PSPLA 1|PSPLA1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53H76
- Q53H76
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together