Anti-NF2 Rabbit Polyclonal Antibody
77832-911
New Product
- Antigen name:Merlin
- Antigen symbol:NF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P35240
- Antigen synonyms:ACN| BANF| Bilateral acoustic neuroma| MERL_HUMAN| Merlin| Moesin ezrin radixin like protein| Moesin ezrin radizin like| Moesin-ezrin-radixin-like protein| Neurofibromatosis 2| Neurofibromatosis type 2| Neurofibromatosis2| Neurofibromin 2| Neurofibromin-2| Neurofibromin2| NF 2| Nf2| SCH| Schwannomerlin| Schwannomin
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: NF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat