Anti-GEMIN2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: GEMIN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-600
- 77799-599
- Antigen symbol:
- GEMIN2
- GEMIN2
- Antigen name:
- Gem-associated protein 2 protein (1-280AA)
- Gem-associated protein 2 protein (1-280AA)
- Antigen synonyms:
- Component of gems 2|Gem (nuclear organelle) associated protein 2|Gem associated protein 2|GEMI2_HUMAN|Gemin-2|gemin2|SIP 1|SIP-1|SIP1|SIP1 delta|SIP1-delta|SMN interacting protein 1|SMN interacting protein 1 delta|SMN-interacting protein 1|Survival interacting protein 1|Survival of motor neuron protein interacting protein 1|Survival of motor neuron protein-interacting protein 1
- Component of gems 2|Gem (nuclear organelle) associated protein 2|Gem associated protein 2|GEMI2_HUMAN|Gemin-2|gemin2|SIP 1|SIP-1|SIP1|SIP1 delta|SIP1-delta|SMN interacting protein 1|SMN interacting protein 1 delta|SMN-interacting protein 1|Survival interacting protein 1|Survival of motor neuron protein interacting protein 1|Survival of motor neuron protein-interacting protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14893
- O14893
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together