Anti-SPRY1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPRY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-929
- 77800-928
- Antigen symbol:
- SPRY1
- SPRY1
- Antigen name:
- Protein sprouty homolog 1 protein (1-178AA)
- Protein sprouty homolog 1 protein (1-178AA)
- Antigen synonyms:
- hSPRY1| Protein sprouty homolog 1| Sprouty 1| Sprouty homolog 1 antagonist of FGF signaling| Sprouty homolog 1| Spry-1| Spry1| SPY1_HUMAN
- hSPRY1| Protein sprouty homolog 1| Sprouty 1| Sprouty homolog 1 antagonist of FGF signaling| Sprouty homolog 1| Spry-1| Spry1| SPY1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43609
- O43609
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together