Anti-CNDP2 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: CNDP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77834-198
- 77834-199
- Antigen symbol:
- CNDP2
- CNDP2
- Antigen name:
- CytosolicNon-specific dipeptidase protein (2-475AA)
- CytosolicNon-specific dipeptidase protein (2-475AA)
- Antigen synonyms:
- 0610010E05Rik|C76600|Carnosinase |tissue|Carnosinase 2|CN 2|CN2|CNDP 2|CNDP dipeptidase 2 (metallopeptidase M20 family)|CNDP dipeptidase 2|CNDP2|CNDP2_HUMAN|CPGL|CytosolicNon specific dipeptidase|CytosolicNon-specific dipeptidase|CytosolicNonspecific dipeptidase|Dip-2|FLJ10830|FLJ35544|Glutamate carboxypeptidase like protein 1|Glutamate carboxypeptidase-like protein 1|HsT2298|Nonspecific dipeptidase|cutosolic|Pep-1|Pep1|PEPA|Peptidase A
- 0610010E05Rik|C76600|Carnosinase |tissue|Carnosinase 2|CN 2|CN2|CNDP 2|CNDP dipeptidase 2 (metallopeptidase M20 family)|CNDP dipeptidase 2|CNDP2|CNDP2_HUMAN|CPGL|CytosolicNon specific dipeptidase|CytosolicNon-specific dipeptidase|CytosolicNonspecific dipeptidase|Dip-2|FLJ10830|FLJ35544|Glutamate carboxypeptidase like protein 1|Glutamate carboxypeptidase-like protein 1|HsT2298|Nonspecific dipeptidase|cutosolic|Pep-1|Pep1|PEPA|Peptidase A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96KP4
- Q96KP4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together