Anti-PKD2 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PKD2
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-461
- 77823-460
- Antigen symbol:
- PKD2
- PKD2
- Antigen name:
- Polycystin-2 protein (680-968AA)
- Polycystin-2 protein (680-968AA)
- Antigen synonyms:
- APKD2|Autosomal dominant polycystic kidney disease type II|Autosomal dominant polycystic kidney disease type II protein|MGC138466|MGC138468|PC 2|PC2|PKD 2|PKD2|PKD2_HUMAN|PKD4|Polycystic kidney disease 2 (autosomal dominant)|Polycystic kidney disease 2|Polycystic kidney disease 2 protein|Polycystin 2|Polycystin 2 transient receptor potential cation channel|Polycystin-2|Polycystin2|Polycystwin|R48321|Transient receptor potential cation channel subfamily P member 2|TRPP2
- APKD2|Autosomal dominant polycystic kidney disease type II|Autosomal dominant polycystic kidney disease type II protein|MGC138466|MGC138468|PC 2|PC2|PKD 2|PKD2|PKD2_HUMAN|PKD4|Polycystic kidney disease 2 (autosomal dominant)|Polycystic kidney disease 2|Polycystic kidney disease 2 protein|Polycystin 2|Polycystin 2 transient receptor potential cation channel|Polycystin-2|Polycystin2|Polycystwin|R48321|Transient receptor potential cation channel subfamily P member 2|TRPP2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13563
- Q13563
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together