Anti-ARL8B Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARL8B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-520
- 77837-519
- Antigen symbol:
- ARL8B
- ARL8B
- Antigen name:
- ADP-ribosylation factor-like protein 8B protein (20-186AA)
- ADP-ribosylation factor-like protein 8B protein (20-186AA)
- Antigen synonyms:
- ADP ribosylation factor like 10C|ADP ribosylation factor like 8B|ADP-ribosylation factor-like protein 10C|ADP-ribosylation factor-like protein 8B|ARL10C|ARL8B_HUMAN|FLJ10702|Gie1|Novel small G protein indispensable for equal chromosome segregation 1
- ADP ribosylation factor like 10C|ADP ribosylation factor like 8B|ADP-ribosylation factor-like protein 10C|ADP-ribosylation factor-like protein 8B|ARL10C|ARL8B_HUMAN|FLJ10702|Gie1|Novel small G protein indispensable for equal chromosome segregation 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVJ2
- Q9NVJ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together