Anti-PRDM16 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PRDM16
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-525
- 77838-524
- Antigen symbol:
- PRDM16
- PRDM16
- Antigen name:
- PR domain zinc finger protein 16 protein (448-683AA)
- PR domain zinc finger protein 16 protein (448-683AA)
- Antigen synonyms:
- CMD1LL|KIAA1675|LVNC8|MDS1/EVI1 like gene 1|MDS1/EVI1-like gene 1|MEL1|PFM 13|PFM13|PR domain containing 16|PR domain containing protein 16|PR domain zinc finger protein 16|PR domain-containing protein 16|PRD16_HUMAN|Prdm16|Transcription factor MEL 1|Transcription factor MEL1
- CMD1LL|KIAA1675|LVNC8|MDS1/EVI1 like gene 1|MDS1/EVI1-like gene 1|MEL1|PFM 13|PFM13|PR domain containing 16|PR domain containing protein 16|PR domain zinc finger protein 16|PR domain-containing protein 16|PRD16_HUMAN|Prdm16|Transcription factor MEL 1|Transcription factor MEL1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HAZ2
- Q9HAZ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together