Anti-d2hgdh Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: d2hgdh
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Specifications
- Catalog No:
- 77817-937
- 77817-938
- Antigen symbol:
- d2hgdh
- d2hgdh
- Antigen name:
- Recombinant Zebrafish D-2-hydroxyglutarate dehydrogenase, mitochondrial protein (56-533AA)
- Recombinant Zebrafish D-2-hydroxyglutarate dehydrogenase, mitochondrial protein (56-533AA)
- Antigen synonyms:
- d2hgdh|zgc:158661D-2-hydroxyglutarate dehydrogenase|mitochondrial|EC 1.1.99.-
- d2hgdh|zgc:158661D-2-hydroxyglutarate dehydrogenase|mitochondrial|EC 1.1.99.-
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Zebrafish
- Zebrafish
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A1L258
- A1L258
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together