Anti-APAF1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: APAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77807-699
- 77807-698
- Antigen symbol:
- APAF1
- APAF1
- Antigen name:
- Synthetic peptide of APAF1
- Synthetic peptide of APAF1
- Antigen synonyms:
- APAF 1|Apaf-1|APAF_HUMAN|Apaf1|Apoptotic peptidase activating factor 1|Apoptotic protease activating factor 1|Apoptotic protease activating factor|Apoptotic protease-activating factor 1|CED 4|CED4|KIAA0413
- APAF 1|Apaf-1|APAF_HUMAN|Apaf1|Apoptotic peptidase activating factor 1|Apoptotic protease activating factor 1|Apoptotic protease activating factor|Apoptotic protease-activating factor 1|CED 4|CED4|KIAA0413
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14727
- O14727
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together