Anti-HSD17B8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: HSD17B8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77821-120
- 77821-119
- Antigen symbol:
- HSD17B8
- HSD17B8
- Antigen name:
- Fusion protein of HSD17B8
- Fusion protein of HSD17B8
- Antigen synonyms:
- 17 beta HSD 8| 17 beta hydroxysteroid dehydrogenase 8| 17-beta-HSD 8| 17-beta-hydroxysteroid dehydrogenase 8| 17-beta-hydroxysteroid dehydrogenase VIII| 3-oxoacyl-[acyl-carrier-protein] reductase| 3-oxoacyl-acyl-carrier protein reductase, E. coli, homolog of| Beta ketoacyl [acyl carrier protein] reductase like| D6S2245E| DHB8_HUMAN| dJ1033B10.9| Estradiol 17 beta dehydrogenase 8| Estradiol 17-beta-dehydrogenase 8| Estrogen 17 oxidoreductase| FABG| FabG, E. coli, homolog of| FabG-like| FABGL| H2 KE6| H2-KE6| HKE6| HSD17B8| Hydroxysteroid (17 beta) dehydrogenase 8| Ke-6| KE6| KE6, mouse homolog of| Protein Ke6| Really interesting new gene 2 protein| RING2| SDR30C1| Short chain dehydrogenase/reductase family 30C member 1| Testosterone 17 beta dehydrogenase 8| Testosterone 17-beta-dehydrogenase 8
- 17 beta HSD 8| 17 beta hydroxysteroid dehydrogenase 8| 17-beta-HSD 8| 17-beta-hydroxysteroid dehydrogenase 8| 17-beta-hydroxysteroid dehydrogenase VIII| 3-oxoacyl-[acyl-carrier-protein] reductase| 3-oxoacyl-acyl-carrier protein reductase, E. coli, homolog of| Beta ketoacyl [acyl carrier protein] reductase like| D6S2245E| DHB8_HUMAN| dJ1033B10.9| Estradiol 17 beta dehydrogenase 8| Estradiol 17-beta-dehydrogenase 8| Estrogen 17 oxidoreductase| FABG| FabG, E. coli, homolog of| FabG-like| FABGL| H2 KE6| H2-KE6| HKE6| HSD17B8| Hydroxysteroid (17 beta) dehydrogenase 8| Ke-6| KE6| KE6, mouse homolog of| Protein Ke6| Really interesting new gene 2 protein| RING2| SDR30C1| Short chain dehydrogenase/reductase family 30C member 1| Testosterone 17 beta dehydrogenase 8| Testosterone 17-beta-dehydrogenase 8
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92506
- Q92506
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together