Anti-RGS3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RGS3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-223
- 77797-224
- Antigen symbol:
- RGS3
- RGS3
- Antigen name:
- Regulator of G-protein signaling 3 protein (1-270AA)
- Regulator of G-protein signaling 3 protein (1-270AA)
- Antigen synonyms:
- C2PA|FLJ20370|FLJ31516|FLJ90496|PDZ RGS 3|PDZ Regulator of G protein signaling 3|Regulator of G protein signalling 3|Regulator of G-protein signaling 3|RGP 3|RGP3|RGS 3|RGS3_HUMAN
- C2PA|FLJ20370|FLJ31516|FLJ90496|PDZ RGS 3|PDZ Regulator of G protein signaling 3|Regulator of G protein signalling 3|Regulator of G-protein signaling 3|RGP 3|RGP3|RGS 3|RGS3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49796
- P49796
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together