Order Entry
Canada
ContactUsLinkComponent
Anti-U2AF1 Rabbit Recombinant Polyclonal Antibody
Anti-U2AF1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-U2AF1 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77811-793
  • 77803-793
  • Antigen symbol:
  • U2AF1
  • U2AF1
  • Antigen name:
  • Splicing factor U2AF 35 kDa subunit protein (1-240AA)
  • Splicing factor U2AF 35 kDa subunit protein (1-240AA)
  • Antigen synonyms:
  • DKFZp313J1712|FP793|Human U2 snRNP auxiliary factor small subunit9|RN|RNU2AF1|Splicing factor U2AF 35 kDa subunit|U2 auxiliary factor 35 kDa subunit|U2 small nuclear ribonucleoprotein auxillary factor 35 KD subunit|U2 small nuclear RNA auxiliary factor 1|U2 snRNP auxiliary factor small subunit|U2(RNU2) small nuclear RNA auxiliary factor 1|U2(RNU2) small nuclear RNA auxiliary factor binding protein|U2AF1|U2AF1_HUMAN|U2AF35|U2AFBP
  • DKFZp313J1712|FP793|Human U2 snRNP auxiliary factor small subunit9|RN|RNU2AF1|Splicing factor U2AF 35 kDa subunit|U2 auxiliary factor 35 kDa subunit|U2 small nuclear ribonucleoprotein auxillary factor 35 KD subunit|U2 small nuclear RNA auxiliary factor 1|U2 snRNP auxiliary factor small subunit|U2(RNU2) small nuclear RNA auxiliary factor 1|U2(RNU2) small nuclear RNA auxiliary factor binding protein|U2AF1|U2AF1_HUMAN|U2AF35|U2AFBP
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q01081
  • Q01081
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together