Order Entry
Canada
ContactUsLinkComponent
Anti-FABP3 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Anti-FABP3 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-FABP3 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77812-146
  • 77812-147
  • Antigen symbol:
  • FABP3
  • FABP3
  • Antigen name:
  • Fatty acid-binding protein, heart protein (2-133AA)
  • Fatty acid-binding protein, heart protein (2-133AA)
  • Antigen synonyms:
  • 422 protein|Cardiac FABP|Cardiac Fatty Acid Binding Protein|FABP 11|FABP 3|FABP11|FABP3|FABPH_HUMAN|Fatty acid binding protein 11|Fatty acid binding protein 3|Fatty acid binding protein 3 muscle and heart|Fatty acid binding protein 3 muscle and heart mammary derived growth inhibitor|Fatty acid binding protein 3 muscle|muscle and heart (mammary derived growth inhibitor)|muscle|Fatty acid binding protein heart|Fatty acid binding protein|heart|muscle and heart|skeletal muscle|Fatty acid-binding protein 3|Fatty acid-binding protein|H FABP|H-FABP|Heart type fatty acid binding protein|Heart-type fatty acid-binding protein|M FABP|M-FABP|Mammary derived growth inhibitor|Mammary-derived growth inhibitor|MDGI|Muscle fatty acid binding protein|Muscle fatty acid-binding protein|Mylein protein P2 homolog|O FABP|OTTHUMP00000003898|P2 adopocyte protein
  • 422 protein|Cardiac FABP|Cardiac Fatty Acid Binding Protein|FABP 11|FABP 3|FABP11|FABP3|FABPH_HUMAN|Fatty acid binding protein 11|Fatty acid binding protein 3|Fatty acid binding protein 3 muscle and heart|Fatty acid binding protein 3 muscle and heart mammary derived growth inhibitor|Fatty acid binding protein 3 muscle|muscle and heart (mammary derived growth inhibitor)|muscle|Fatty acid binding protein heart|Fatty acid binding protein|heart|muscle and heart|skeletal muscle|Fatty acid-binding protein 3|Fatty acid-binding protein|H FABP|H-FABP|Heart type fatty acid binding protein|Heart-type fatty acid-binding protein|M FABP|M-FABP|Mammary derived growth inhibitor|Mammary-derived growth inhibitor|MDGI|Muscle fatty acid binding protein|Muscle fatty acid-binding protein|Mylein protein P2 homolog|O FABP|OTTHUMP00000003898|P2 adopocyte protein
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P05413
  • P05413
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together