Anti-CLU Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CLU
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77809-405
- 77809-406
- Antigen symbol:
- CLU
- CLU
- Antigen name:
- Clusterin protein (23-224AA)
- Clusterin protein (23-224AA)
- Antigen synonyms:
- 40|AAG 4|AAG4|Aging associated protein 4|Aging-associated gene 4 protein|AI893575|APO J|Apo-J|APOJ|ApoJalpha|ApoJbeta|Apolipoprotein J|ApolipoproteinJ|CLI|CLU|CLU1|CLU2|CLUS_HUMAN|Clusterin alpha chain|Clusterin|Clusterin beta chain|Complement associated protein SP 40 40|Complement associated protein SP 40|Complement associated protein SP40|Complement cytolysis inhibitor a chain|Complement cytolysis inhibitor|Complement cytolysis inhibitor b chain|Complement lysis inhibitor|Complement-associated protein SP-40|D14Ucla3|Dimeric acid glycoprotein|Glycoprotein 80|Glycoprotein III|GP80|Ku70-binding protein 1|KUB 1|KUB1|MGC24903|NA1/NA2|RATTRPM2B|SGP 2|SGP2|SP 40|SP40|Sugp-2|Sulfated glycoprotein 2|Testosterone repressed prostate message 2|Testosterone-repressed prostate message 2|TRPM 2|TRPM-2|TRPM2|TRPM2B|Trpmb
- 40|AAG 4|AAG4|Aging associated protein 4|Aging-associated gene 4 protein|AI893575|APO J|Apo-J|APOJ|ApoJalpha|ApoJbeta|Apolipoprotein J|ApolipoproteinJ|CLI|CLU|CLU1|CLU2|CLUS_HUMAN|Clusterin alpha chain|Clusterin|Clusterin beta chain|Complement associated protein SP 40 40|Complement associated protein SP 40|Complement associated protein SP40|Complement cytolysis inhibitor a chain|Complement cytolysis inhibitor|Complement cytolysis inhibitor b chain|Complement lysis inhibitor|Complement-associated protein SP-40|D14Ucla3|Dimeric acid glycoprotein|Glycoprotein 80|Glycoprotein III|GP80|Ku70-binding protein 1|KUB 1|KUB1|MGC24903|NA1/NA2|RATTRPM2B|SGP 2|SGP2|SP 40|SP40|Sugp-2|Sulfated glycoprotein 2|Testosterone repressed prostate message 2|Testosterone-repressed prostate message 2|TRPM 2|TRPM-2|TRPM2|TRPM2B|Trpmb
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10909
- P10909
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together