Anti-SLC7A2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SLC7A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77800-158
- 77800-159
- Antigen symbol:
- SLC7A2
- SLC7A2
- Antigen name:
- Cationic amino acid transporter 2 protein (604-658AA)
- Cationic amino acid transporter 2 protein (604-658AA)
- Antigen synonyms:
- ATRC2|20.5|Amino acid transporter|cationic 2 (low affinity)|ATRC 2|CAT 2|CAT-2|CAT2|Cat2a|Cat2b|Cationic amino acid transporter 2A|cationic amino acid transporter|y+ system|low affinity cationic amino acid transporter 2|CTR2_HUMAN|HCAT 2|HCAT2|Low affinity cationic amino acid transporter 2|OTTHUMP00000216432|OTTHUMP00000216433|OTTMUSP00000041667|OTTMUSP00000041669|RCAT2|SLC7A 2|Solute carrier family 7 member 2|T cell early activation protein|Tea
- ATRC2|20.5|Amino acid transporter|cationic 2 (low affinity)|ATRC 2|CAT 2|CAT-2|CAT2|Cat2a|Cat2b|Cationic amino acid transporter 2A|cationic amino acid transporter|y+ system|low affinity cationic amino acid transporter 2|CTR2_HUMAN|HCAT 2|HCAT2|Low affinity cationic amino acid transporter 2|OTTHUMP00000216432|OTTHUMP00000216433|OTTMUSP00000041667|OTTMUSP00000041669|RCAT2|SLC7A 2|Solute carrier family 7 member 2|T cell early activation protein|Tea
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52569
- P52569
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together