Anti-GBA2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GBA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-811
- 77840-812
- Antigen symbol:
- GBA2
- GBA2
- Antigen name:
- Non-lysosomal glucosylceramidase protein (656-867AA)
- Non-lysosomal glucosylceramidase protein (656-867AA)
- Antigen synonyms:
- GBA2|KIAA1605|SPG46|AD035Non-lysosomal glucosylceramidase|NLGase|EC 3.2.1.45|Beta-glucocerebrosidase 2|Beta-glucosidase 2|Bile acid beta-glucosidase GBA2|Bile acid glucosyl transferase GBA2|Cholesterol glucosyltransferase GBA2|EC 2.4.1.-|Cholesteryl-beta-glucosidase GBA2|EC 3.2.1.104|Glucosylceramidase 2
- GBA2|KIAA1605|SPG46|AD035Non-lysosomal glucosylceramidase|NLGase|EC 3.2.1.45|Beta-glucocerebrosidase 2|Beta-glucosidase 2|Bile acid beta-glucosidase GBA2|Bile acid glucosyl transferase GBA2|Cholesterol glucosyltransferase GBA2|EC 2.4.1.-|Cholesteryl-beta-glucosidase GBA2|EC 3.2.1.104|Glucosylceramidase 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCG7
- Q9HCG7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together