Anti-EDEM2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EDEM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-265
- 77835-266
- Antigen symbol:
- EDEM2
- EDEM2
- Antigen name:
- ER degradation-enhancing alpha-mannosidase-like protein 2 protein (22-300AA)
- ER degradation-enhancing alpha-mannosidase-like protein 2 protein (22-300AA)
- Antigen synonyms:
- bA4204.1|C20orf31|C20orf49|EDEM 2|EDEM2|EDEM2_HUMAN|ER degradation enhancer mannosidase alpha like 2|ER degradation enhancing alpha mannosidase like 2|ER degradation enhancing alpha mannosidase like protein 2|ER degradation enhancing mannosidase like protein 2|ER degradation-enhancing alpha-mannosidase-like 2|FLJ10783|UNQ573/PRO1135
- bA4204.1|C20orf31|C20orf49|EDEM 2|EDEM2|EDEM2_HUMAN|ER degradation enhancer mannosidase alpha like 2|ER degradation enhancing alpha mannosidase like 2|ER degradation enhancing alpha mannosidase like protein 2|ER degradation enhancing mannosidase like protein 2|ER degradation-enhancing alpha-mannosidase-like 2|FLJ10783|UNQ573/PRO1135
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BV94
- Q9BV94
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together