Anti-TNF Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: TNF
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Specifications
- Catalog No:
- 77812-947
- 77812-948
- Antigen symbol:
- TNF
- TNF
- Antigen name:
- Recombinant Bovine Tumor necrosis factor protein (78-234AA)
- Recombinant Bovine Tumor necrosis factor protein (78-234AA)
- Antigen synonyms:
- TNF| TNFA| TNFSF2| Tumor necrosis factor| Cachectin| TNF-alpha| Tumor necrosis factor ligand superfamily member 2| TNF-a) [Cleaved into: Tumor necrosis factor| membrane form| N-terminal fragment| NTF)| Intracellular domain 1| ICD1)| Intracellular domain 2| ICD2)| C-domain 1| C-domain 2| soluble form]
- TNF| TNFA| TNFSF2| Tumor necrosis factor| Cachectin| TNF-alpha| Tumor necrosis factor ligand superfamily member 2| TNF-a) [Cleaved into: Tumor necrosis factor| membrane form| N-terminal fragment| NTF)| Intracellular domain 1| ICD1)| Intracellular domain 2| ICD2)| C-domain 1| C-domain 2| soluble form]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Bovine
- Bovine
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q06599
- Q06599
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together