Anti-SYF2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SYF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-682
- 77801-681
- Antigen symbol:
- SYF2
- SYF2
- Antigen name:
- Pre-mRNA-splicing factor SYF2 protein (2-243AA)
- Pre-mRNA-splicing factor SYF2 protein (2-243AA)
- Antigen synonyms:
- CBPIN|CCNDBP1 interactor|CCNDBP1-interactor|DKFZp564O2082|fSAP29|Functional spliceosome associated protein 29|GCIP interacting protein p29|GCIPIP|NTC31|P29|Pre mRNA splicing factor SYF2|Pre-mRNA-splicing factor syf2|syf2|SYF2 homolog|SYF2 homolog RNA splicing factor (S. cerevisiae)|SYF2 homolog RNA splicing factor|SYF2_HUMAN
- CBPIN|CCNDBP1 interactor|CCNDBP1-interactor|DKFZp564O2082|fSAP29|Functional spliceosome associated protein 29|GCIP interacting protein p29|GCIPIP|NTC31|P29|Pre mRNA splicing factor SYF2|Pre-mRNA-splicing factor syf2|syf2|SYF2 homolog|SYF2 homolog RNA splicing factor (S. cerevisiae)|SYF2 homolog RNA splicing factor|SYF2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95926
- O95926
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together