Anti-ptxA Rabbit Recombinant Polyclonal Antibody
Catalog # 77816-964
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Recombinant Bordetella pertussis Pertussis toxin subunit 1 protein (35-269AA)
- Antigen symbol:ptxA
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:B. pertussis
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P04977
- Antigen synonyms:ptxA|BP3783|Pertussis toxin subunit 1|PTX S1|Islet-activating protein S1|IAP S1|NAD-dependent ADP-ribosyltransferase|EC 2.4.2.-
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: ptxA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: B. pertussis