Anti-SLC14A1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SLC14A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77822-879
- 77822-880
- Antigen symbol:
- SLC14A1
- SLC14A1
- Antigen name:
- Urea transporter 1 protein (1-51AA)
- Urea transporter 1 protein (1-51AA)
- Antigen synonyms:
- blood group Kidd urea transporter| erythrocyte| HsT1341| HUT11| JK| Kidd| kidd (JK) blood group urea transporter B1| RACH1| Slc14a1| solute carrier family 14 (urea transporter) member 1 (Kidd blood group)| Solute carrier family 14 member 1| Urea transporter 1| Urea transporter| urea transporter B1| Urea transporter erythrocyte| urea transporter JK glycoprotein| UT B1| UT1| UT1_HUMAN| UTE
- blood group Kidd urea transporter| erythrocyte| HsT1341| HUT11| JK| Kidd| kidd (JK) blood group urea transporter B1| RACH1| Slc14a1| solute carrier family 14 (urea transporter) member 1 (Kidd blood group)| Solute carrier family 14 member 1| Urea transporter 1| Urea transporter| urea transporter B1| Urea transporter erythrocyte| urea transporter JK glycoprotein| UT B1| UT1| UT1_HUMAN| UTE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13336
- Q13336
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together