Anti-RASSF7 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RASSF7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-992
- 77835-993
- Antigen symbol:
- RASSF7
- RASSF7
- Antigen name:
- Ras association domain-containing protein 7 protein (300-368AA)
- Ras association domain-containing protein 7 protein (300-368AA)
- Antigen synonyms:
- C11orf13|Chromosome 11 open reading frame 13|HRAS1|HRAS1 related cluster 1|HRAS1 related cluster protein 1|HRAS1-related cluster protein 1|HRC1|MGC126069|MGC126070|Ras association (RalGDS/AF 6) domain family (N terminal) member 7|Ras association (RalGDS/AF 6) domain family 7|Ras association domain containing protein 7|Ras association domain-containing protein 7|RASF7_HUMAN|RASSF 7|RASSF7
- C11orf13|Chromosome 11 open reading frame 13|HRAS1|HRAS1 related cluster 1|HRAS1 related cluster protein 1|HRAS1-related cluster protein 1|HRC1|MGC126069|MGC126070|Ras association (RalGDS/AF 6) domain family (N terminal) member 7|Ras association (RalGDS/AF 6) domain family 7|Ras association domain containing protein 7|Ras association domain-containing protein 7|RASF7_HUMAN|RASSF 7|RASSF7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02833
- Q02833
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together