Anti-RECQL4 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: RECQL4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77796-972
- 77796-973
- Antigen symbol:
- RECQL4
- RECQL4
- Antigen name:
- ATP-dependent DNA helicase Q4 protein (200-337AA)
- ATP-dependent DNA helicase Q4 protein (200-337AA)
- Antigen synonyms:
- ATP dependent DNA helicase Q4|ATP-dependent DNA helicase Q4|DNA helicase|DNA helicase, RecQ like type 4|RECQ 4|RECQ L4|RecQ protein 4|RecQ protein like 4|RecQ protein-like 4|RecQ-like type 4|RecQ4|RECQ4_HUMAN|RECQL 4|RECQL4|RTS
- ATP dependent DNA helicase Q4|ATP-dependent DNA helicase Q4|DNA helicase|DNA helicase, RecQ like type 4|RECQ 4|RECQ L4|RecQ protein 4|RecQ protein like 4|RecQ protein-like 4|RecQ-like type 4|RecQ4|RECQ4_HUMAN|RECQL 4|RECQL4|RTS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94761
- O94761
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together