Anti-POMK Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POMK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-187
- 77836-186
- Antigen symbol:
- POMK
- POMK
- Antigen name:
- Protein O-mannose kinase protein (44-350AA)
- Protein O-mannose kinase protein (44-350AA)
- Antigen synonyms:
- FLJ23356|MDDGA12||Probable inactive protein kinase-like protein SgK196|Protein kinase like protein SgK196|Protein kinase-like protein SgK196|Protein O-mannose kinase|SG196_HUMAN|SGK196|Sugen kinase 196
- FLJ23356|MDDGA12||Probable inactive protein kinase-like protein SgK196|Protein kinase like protein SgK196|Protein kinase-like protein SgK196|Protein O-mannose kinase|SG196_HUMAN|SGK196|Sugen kinase 196
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H5K3
- Q9H5K3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together