Anti-GINS1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GINS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-105
- 77823-104
- Antigen symbol:
- GINS1
- GINS1
- Antigen name:
- DNA replication complex GINS protein PSF1 protein (1-196AA)
- DNA replication complex GINS protein PSF1 protein (1-196AA)
- Antigen synonyms:
- DNA replication complex GINS protein PSF1|GINS complex subunit 1 (Psf1 homolog)|GINS complex subunit 1|GINS1|partner of sld five-1|partner of sld5|PSF1 homolog|PSF1_HUMAN|RP4-691N24.2
- DNA replication complex GINS protein PSF1|GINS complex subunit 1 (Psf1 homolog)|GINS complex subunit 1|GINS1|partner of sld five-1|partner of sld5|PSF1 homolog|PSF1_HUMAN|RP4-691N24.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14691
- Q14691
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together