Order Entry
Canada
ContactUsLinkComponent
Anti-PRKD3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-PRKD3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-PRKD3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77795-391
  • 77795-392
  • Antigen symbol:
  • PRKD3
  • PRKD3
  • Antigen name:
  • Serine/threonine-protein kinase D3 protein (326-510AA)
  • Serine/threonine-protein kinase D3 protein (326-510AA)
  • Antigen synonyms:
  • EPK 2|EPK2|KPCD3_HUMAN|nPKC nu|nPKC-nu|nPKCnu|nu|PKCnu|PKD 3|PKD3|PRK D3|PRKCN|PRKD 3|Prkd3|Protein kinase C|Protein kinase C nu|Protein kinase C nu type|Protein kinase D3|Protein kinase EPK 2|Protein kinase EPK2|Serine threonine protein kinase|Serine/threonine protein kinase D3|Serine/threonine-protein kinase D3
  • EPK 2|EPK2|KPCD3_HUMAN|nPKC nu|nPKC-nu|nPKCnu|nu|PKCnu|PKD 3|PKD3|PRK D3|PRKCN|PRKD 3|Prkd3|Protein kinase C|Protein kinase C nu|Protein kinase C nu type|Protein kinase D3|Protein kinase EPK 2|Protein kinase EPK2|Serine threonine protein kinase|Serine/threonine protein kinase D3|Serine/threonine-protein kinase D3
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O94806
  • O94806
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together