Anti-TRIM3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-542
- 77803-542
- Antigen symbol:
- TRIM3
- TRIM3
- Antigen name:
- Tripartite motif-containing protein 3 protein (285-487AA)
- Tripartite motif-containing protein 3 protein (285-487AA)
- Antigen synonyms:
- BERP|Brain expressed ring finger|Brain-expressed RING finger protein|FLJ16135|HAC1|RING finger protein 22|RING finger protein 97|RNF22|RNF97|Trim3|TRIM3_HUMAN|Tripartite motif containing 3|Tripartite motif protein TRIM3|Tripartite motif-containing protein 3
- BERP|Brain expressed ring finger|Brain-expressed RING finger protein|FLJ16135|HAC1|RING finger protein 22|RING finger protein 97|RNF22|RNF97|Trim3|TRIM3_HUMAN|Tripartite motif containing 3|Tripartite motif protein TRIM3|Tripartite motif-containing protein 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75382
- O75382
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together