Anti-STX4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: STX4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-141
- 77825-142
- Antigen symbol:
- STX4
- STX4
- Antigen name:
- Syntaxin-4 protein (1-275AA)
- Syntaxin-4 protein (1-275AA)
- Antigen synonyms:
- NY REN 31 antigen|p35 2|PLACENTAL|Renal carcinoma antigen NY REN 31|Renal carcinoma antigen NY-REN-31|Stx4|STX4_HUMAN|STX4A|Syntaxin 4|syntaxin 4A (placental)|Syntaxin 4A|Syntaxin-4
- NY REN 31 antigen|p35 2|PLACENTAL|Renal carcinoma antigen NY REN 31|Renal carcinoma antigen NY-REN-31|Stx4|STX4_HUMAN|STX4A|Syntaxin 4|syntaxin 4A (placental)|Syntaxin 4A|Syntaxin-4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12846
- Q12846
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together