Anti-PRPF4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: PRPF4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-572
- 77795-571
- Antigen symbol:
- PRPF4
- PRPF4
- Antigen name:
- U4/U6 small nuclear ribonucleoprotein Prp4 protein (1-260AA)
- U4/U6 small nuclear ribonucleoprotein Prp4 protein (1-260AA)
- Antigen synonyms:
- hPrp4|HPRP4P|PRP4|PRP4 homolog|PRP4 pre mRNA processing factor 4 homolog (yeast)|PRP4/STK/WD splicing factor|PRP4_HUMAN|Prp4p|PRPF4|RP70|SNRNP60|U4/U6 small nuclear ribonucleoprotein Prp4|U4/U6 snRNP 60 kDa protein|WD splicing factor Prp4
- hPrp4|HPRP4P|PRP4|PRP4 homolog|PRP4 pre mRNA processing factor 4 homolog (yeast)|PRP4/STK/WD splicing factor|PRP4_HUMAN|Prp4p|PRPF4|RP70|SNRNP60|U4/U6 small nuclear ribonucleoprotein Prp4|U4/U6 snRNP 60 kDa protein|WD splicing factor Prp4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43172
- O43172
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together