Anti-WNK3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WNK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-324
- 77835-325
- Antigen symbol:
- WNK3
- WNK3
- Antigen name:
- Serine/threonine-protein kinase WNK3 protein (1018-1181AA)
- Serine/threonine-protein kinase WNK3 protein (1018-1181AA)
- Antigen synonyms:
- KIAA1566|PRKWNK 3|PRKWNK3|Protein kinase lysine deficient 3|Protein kinase lysine-deficient 3|Protein kinase with no lysine 3|Serine/threonine protein kinase WNK3|Serine/threonine-protein kinase WNK3|WNK 3|WNK lysine deficient protein kinase 3|Wnk3|WNK3_HUMAN
- KIAA1566|PRKWNK 3|PRKWNK3|Protein kinase lysine deficient 3|Protein kinase lysine-deficient 3|Protein kinase with no lysine 3|Serine/threonine protein kinase WNK3|Serine/threonine-protein kinase WNK3|WNK 3|WNK lysine deficient protein kinase 3|Wnk3|WNK3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BYP7
- Q9BYP7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together