Order Entry
Canada
ContactUsLinkComponent
Anti-ECH1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-ECH1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ECH1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77824-271
  • 77824-270
  • Antigen symbol:
  • ECH1
  • ECH1
  • Antigen name:
  • Delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial protein (36-135AA)
  • Delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial protein (36-135AA)
  • Antigen synonyms:
  • 4)-dienoyl-CoA isomerase|5)-Delta(2|Delta(3 5) Delta(2 4) dienoyl CoA isomerase|Delta(3 5) Delta(2 4) dienoyl CoA isomerase mitochondrial|Delta(3|Delta3 5 delta2 4 dienoyl CoA isomerase|Dienoyl CoA isomerase|ECH 1|Ech1|ECH1_HUMAN|Enoyl Coenzyme A hydratase 1|Enoyl Coenzyme A hydratase 1 peroxisomal|HPXEL|mitochondrial|MS1034|Peroxisomal enoyl CoA hydratase 1|Peroxisomal enoyl coenzyme A hydratase like protein
  • 4)-dienoyl-CoA isomerase|5)-Delta(2|Delta(3 5) Delta(2 4) dienoyl CoA isomerase|Delta(3 5) Delta(2 4) dienoyl CoA isomerase mitochondrial|Delta(3|Delta3 5 delta2 4 dienoyl CoA isomerase|Dienoyl CoA isomerase|ECH 1|Ech1|ECH1_HUMAN|Enoyl Coenzyme A hydratase 1|Enoyl Coenzyme A hydratase 1 peroxisomal|HPXEL|mitochondrial|MS1034|Peroxisomal enoyl CoA hydratase 1|Peroxisomal enoyl coenzyme A hydratase like protein
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q13011
  • Q13011
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, protein G purified
  • >95%, protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together