Anti-POLD3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-153
- 77823-152
- Antigen symbol:
- POLD3
- POLD3
- Antigen name:
- DNA polymerase delta subunit 3 protein (203-275AA)
- DNA polymerase delta subunit 3 protein (203-275AA)
- Antigen synonyms:
- DNA polymerase delta subunit 3|DNA polymerase delta subunit p66|DNA polymerase delta|subunit 3|DNA polymerase subunit delta p66|DPOD3_HUMAN|KIAA0039|P66|P68|Pol delta C subunit (p66)|Pold3|Polymerase (DNA directed)|delta 3|Polymerase (DNA-directed)|accessory subunit|PPP1R128|Protein phosphatase 1|regulatory subunit 128
- DNA polymerase delta subunit 3|DNA polymerase delta subunit p66|DNA polymerase delta|subunit 3|DNA polymerase subunit delta p66|DPOD3_HUMAN|KIAA0039|P66|P68|Pol delta C subunit (p66)|Pold3|Polymerase (DNA directed)|delta 3|Polymerase (DNA-directed)|accessory subunit|PPP1R128|Protein phosphatase 1|regulatory subunit 128
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15054
- Q15054
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together