Anti-MAN2A1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAN2A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-243
- 77822-242
- Antigen symbol:
- MAN2A1
- MAN2A1
- Antigen name:
- Alpha-mannosidase 2 protein (31-181AA)
- Alpha-mannosidase 2 protein (31-181AA)
- Antigen synonyms:
- 3-1|6-alpha-mannosidase|Alpha mannosidase 2|alpha Mannosidase II|Alpha-mannosidase 2|AMAN II|Golgi alpha mannosidase II|Golgi alpha-mannosidase II|Golgi integral membrane protein 7|GOLIM7|MA2A1_HUMAN|MAN II|Man2a1|MANA 2|MANA2|MANII|Mann II|Mannosidase alpha class 2A member 1|Mannosidase Two|Mannosidase|alpha type II|alpha|II|Mannosyl oligosaccharide 1 3 1 6 alpha mannosidase|Mannosyl oligosaccharide 1,3 1,6 alpha mannosidase|Mannosyl-oligosaccharide 1
- 3-1|6-alpha-mannosidase|Alpha mannosidase 2|alpha Mannosidase II|Alpha-mannosidase 2|AMAN II|Golgi alpha mannosidase II|Golgi alpha-mannosidase II|Golgi integral membrane protein 7|GOLIM7|MA2A1_HUMAN|MAN II|Man2a1|MANA 2|MANA2|MANII|Mann II|Mannosidase alpha class 2A member 1|Mannosidase Two|Mannosidase|alpha type II|alpha|II|Mannosyl oligosaccharide 1 3 1 6 alpha mannosidase|Mannosyl oligosaccharide 1,3 1,6 alpha mannosidase|Mannosyl-oligosaccharide 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16706
- Q16706
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together