Anti-ADCY1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: ADCY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77821-512
- 77821-513
- Antigen symbol:
- ADCY1
- ADCY1
- Antigen name:
- Adenylate cyclase type 1 protein (416-610AA)
- Adenylate cyclase type 1 protein (416-610AA)
- Antigen synonyms:
- Adenylate cyclase 1|fetal brain|type I|3',5'-cyclic AMP synthetase|AC1|Adcy1|ADCY1_HUMAN|adenyl cyclase|adenylate cyclase 1 (brain)|Adenylate cyclase type 1|Adenylate cyclase type I|Adenylyl cyclase 1|ATP pyrophosphate lyase|ATP pyrophosphate-lyase 1|Brain adenylate cyclase 1|Ca(2+)/calmodulin-activated adenylyl cyclase|deafness|autosomal recessive 44|DFNB44
- Adenylate cyclase 1|fetal brain|type I|3',5'-cyclic AMP synthetase|AC1|Adcy1|ADCY1_HUMAN|adenyl cyclase|adenylate cyclase 1 (brain)|Adenylate cyclase type 1|Adenylate cyclase type I|Adenylyl cyclase 1|ATP pyrophosphate lyase|ATP pyrophosphate-lyase 1|Brain adenylate cyclase 1|Ca(2+)/calmodulin-activated adenylyl cyclase|deafness|autosomal recessive 44|DFNB44
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q08828
- Q08828
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together